Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S172

Sigma-Aldrich

Sarafotoxin S6c

Synonym(s):

SRTX-c

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C103H147N27O37S5
CAS Number:
Molecular Weight:
2515.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

technique(s)

ligand binding assay: suitable

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(NC(=O)C(CC(O)=O)NC(=O)C(CCC(N)=O)NC(=O)C(Cc1cnc[nH]1)NC(=O)C2CSSCC(N)C(=O)NC(C(C)O)C(=O)NC3CSSCC(NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(=O)C(NC(=O)C(CCSC)NC(=O)C(CC(O)=O)NC(=O)C(CC(N)=O)NC3=O)C(C)O)C(=O)NC(CC(C)C)C(=O)NC(CC(N)=O)C(=O)NC(Cc4ccccc4)C(=O)N2)C(C)C)C(=O)NC(Cc5c[nH]c6ccccc56)C(O)=O

InChI

1S/C103H147N27O37S5/c1-10-46(6)80(100(163)123-67(103(166)167)30-50-37-109-54-19-15-14-18-52(50)54)128-99(162)79(45(4)5)127-95(158)66(36-78(144)145)120-85(148)55(20-23-71(105)133)112-90(153)61(31-51-38-108-43-110-51)117-97(160)69-40-170-169-39-53(104)83(146)129-81(47(7)131)102(165)126-70-42-172-171-41-68(96(159)115-59(28-44(2)3)88(151)118-62(32-72(106)134)91(154)116-60(89(152)125-69)29-49-16-12-11-13-17-49)124-86(149)57(22-25-75(138)139)111-84(147)56(21-24-74(136)137)113-94(157)65(35-77(142)143)122-101(164)82(48(8)132)130-87(150)58(26-27-168-9)114-93(156)64(34-76(140)141)121-92(155)63(33-73(107)135)119-98(70)161/h11-19,37-38,43-48,53,55-70,79-82,109,131-132H,10,20-36,39-42,104H2,1-9H3,(H2,105,133)(H2,106,134)(H2,107,135)(H,108,110)(H,111,147)(H,112,153)(H,113,157)(H,114,156)(H,115,159)(H,116,154)(H,117,160)(H,118,151)(H,119,161)(H,120,148)(H,121,155)(H,122,164)(H,123,163)(H,124,149)(H,125,152)(H,126,165)(H,127,158)(H,128,162)(H,129,146)(H,130,150)(H,136,137)(H,138,139)(H,140,141)(H,142,143)(H,144,145)(H,166,167)

InChI key

LXPHPKVWHQLBBA-UHFFFAOYSA-N

Gene Information

human ... OXT(5020)

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service