Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

RAB0360

Sigma-Aldrich

Human MIF ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 8.23-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MIF(4282)

General description

The Human MIF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MIF in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human MIF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahar Mohamed Yehya Elbaradie et al.
BMC women's health, 20(1), 189-189 (2020-09-05)
Endometriosis is a long-standing progressive disease that affects women of reproductive age. Macrophage migration inhibitory factor (MIF) is one of non-invasive blood biomarker that was detected in sera of endometriotic patients. The present study aimed to determine the accuracy of
Hyun Ah Oh et al.
Cells, 9(6) (2020-06-03)
Conventional therapeutic applications of mesenchymal stromal cells (MSCs) focus on cell replacement and differentiation; however, increasing evidence suggests that most of their therapeutic effects are carried out by their various secretions. This study investigated the application of conditioned medium (CM)
Xinxin Wang et al.
Clinical laboratory, 66(11) (2020-11-13)
The current study aims to investigate the clinical significance of serum macrophage migration inhib-itory factor (MIF) and C-C motif chemokine living 23 (CCL23) in patients with acute cerebral infarction (ACI). Seventy-nine patients with ACI were selected and divided into three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service