Skip to Content
MilliporeSigma
All Photos(1)

Documents

PZ0349

Sigma-Aldrich

PF-04449613

≥98% (HPLC)

Synonym(s):

(R)-6-(1-(3-Phenoxyazetidin-1-yl)ethyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4(5H)-one, 1,5-Dihydro-6-[(1R)-1-(3-phenoxy-1-azetidinyl)ethyl]-1-(tetrahydro-2H-pyran-4-yl)-4H-pyrazolo[3,4-d]pyrimidin-4-one, 6-[(1R)-1-(3-Phenoxyazetidin-1-yl)ethyl]-1-(tetrahydro-2H-pyran-4-yl)-1,5-dihydro-4H-pyrazolo[3,4-d]pyrimidin-4-one, PF 4449613, PF-9613

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25N5O3
CAS Number:
Molecular Weight:
395.45
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

PF-04449613 is a potent and selective cGMP-competitive PDE9A inhibitor (IC50 = 24 nM; IC50 >800 nM against other PDE members), displaying more than 1000-fold selectivity over the majority of 79 non-PDE targets investigated with the exception of cytochrome P450 2C19 (IC50 = 1600 nM), dopamine transporter (Ki = 110 nM), μ-opioid receptor (Ki = 3500 nM), and sodium channel binding site 2 (Ki = 470 nM). PF-4449613 is shown to effectively increase cGMP levels in brain (1-10 mg/kg s.c.; mice) and CSF (1-32 mg/kg s.c.; rats and cynomolgus macaques) in vivo with good brain penetration (brain/plasma ratio of 0.8 in mice). PF-04449613 is effective against D-amphetamine-induced auditory gating deficit (by 43% with 1 mg/kg PF-04449613 s.c.; 1 mg/kg AMP i.v.) and transverse aortic constriction-caused cardiac dysfunction (30 mg/kg/12 h p.o.) in mice in a NOS-independent manner.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle M Claffey et al.
Journal of medicinal chemistry, 55(21), 9055-9068 (2012-10-03)
Phosphodiesterase 9A inhibitors have shown activity in preclinical models of cognition with potential application as novel therapies for treating Alzheimer's disease. Our clinical candidate, PF-04447943 (2), demonstrated acceptable CNS permeability in rats with modest asymmetry between central and peripheral compartments
Robin J Kleiman et al.
The Journal of pharmacology and experimental therapeutics, 341(2), 396-409 (2012-02-14)
Cyclic nucleotides are critical regulators of synaptic plasticity and participate in requisite signaling cascades implicated across multiple neurotransmitter systems. Phosphodiesterase 9A (PDE9A) is a high-affinity, cGMP-specific enzyme widely expressed in the rodent central nervous system. In the current study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service