Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P9125

Sigma-Aldrich

Methyl petroselinate

≥99%, liquid

Synonym(s):

Methyl cis-6-octadecenoate, Petroselinic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
Beilstein:
1727035
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

synthetic (organic)

Assay

≥99%

form

liquid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCC\C=C\CCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h13-14H,3-12,15-18H2,1-2H3/b14-13+

InChI key

QRTVDKVXAFJVRU-BUHFOSPRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choyce A Weatherly et al.
Journal of agricultural and food chemistry, 64(6), 1422-1432 (2016-02-09)
Four ionic liquid (IL) columns, SLB-IL59, SLB-IL60, SLB-IL65, and SLB-IL111, were evaluated for more rapid analysis or improved resolution of long-chain methyl and ethyl esters of omega-3, omega-6, and additional positional isomeric and stereoisomeric blends of fatty acids found in
Michal Korinek et al.
Frontiers in pharmacology, 8, 356-356 (2017-07-05)
Increasing prevalence of allergic diseases with an inadequate variety of treatment drives forward search for new alternative drugs. Fatty acids, abundant in nature, are regarded as important bioactive compounds and powerful nutrients playing an important role in lipid homeostasis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service