Skip to Content
MilliporeSigma
All Photos(1)

Documents

P8627

Sigma-Aldrich

D(−)3-Phosphoglyceric acid barium salt

~95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O7PBa
CAS Number:
Molecular Weight:
321.37
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

~95%

storage temp.

−20°C

SMILES string

O.[Ba].O[C@H](COP(O)(O)=O)C(O)=O

InChI

1S/C3H7O7P.Ba.H2O.2H/c4-2(3(5)6)1-10-11(7,8)9;;;;/h2,4H,1H2,(H,5,6)(H2,7,8,9);;1H2;;/t2-;;;;/m1..../s1

InChI key

WKDIHXKLDDUHFS-NNNLZWLESA-N

comparable product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service