Skip to Content
MilliporeSigma
All Photos(1)

Documents

P7061

Sigma-Aldrich

Pluronic® F-68

suitable for plant cell culture, BioReagent

Synonym(s):

Polyoxyethylene-polyoxypropylene block copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H6O.C2H4O)x
CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

description

Contains 100 ppm BHT
non-ionic

product line

BioReagent

mol wt

average mol wt 8350

technique(s)

cell culture | plant: suitable

CMC

0.04 mM (20-25°C)

HLB

29

application(s)

agriculture

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pluronic F-68, a poloxamer, has been used as a supplement to Murashige and Skoog medium to enhance shoot regeneration in citrus rootstock and as a component of plant cell cryopreservation media.

Legal Information

Pluronic is a registered trademark of BASF

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G O Cancino et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 29(4), 317-324 (2001-08-10)
The effects have been studied in vitro of the non-ionic, co-polymer surfactant, Pluronic F-68, on shoot regeneration and bud induction in epicotyl and cotyledon explants of Citrus depressa, a potential alternative rootstock to C. jambhiri for commercial Citrus. Supplementation of
M A Zhao et al.
Plant cell reports, 24(8), 477-481 (2005-05-03)
Cryopreservation has been recognized as a practical and efficient tool for long-term storage of vegetatively propagated plants. This study was conducted to investigate effects of modified vitrification techniques on cryopreservation of potato. In vitro plants of potato cultivars "Superior" and
K C Lowe et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 29(4), 297-316 (2001-08-10)
The storage of prokaryotic and eukaryotic cells at ultra-low temperature in liquid nitrogen (-196 degrees C) is a procedure that has assumed an increasingly important role in underpinning many aspects of biotechnology. For eukaryotic cells, the transition from a cryopreserved
Ola Tuvesson et al.
Cytotechnology, 56(2), 123-136 (2008-11-13)
We have developed a generic transient transfection process at 100 L scale, using HEK293-EBNA cells and PEI as the transfection reagent for the production of recombinant IgG. The process, including large-scale plasmid preparation, expression at bioreactor scale, capture, purification and
Tsz Chung Lai et al.
Colloids and surfaces. B, Biointerfaces, 99, 27-37 (2011-10-18)
Polyplex particles formed with plasmid DNA (pDNA) and Pluronic P85-block-poly{N-[N-(2-aminoethyl)-2-aminoethyl]aspartamide} (P85-b-P[Asp(DET)]) demonstrated highly effective transfection ability compared to PEG-based block cationomer, PEG-b-P[Asp(DET)]. Ternary polyplexes comprising PEG-b-P[Asp(DET)], poly(ethylene oxide)-b-poly(propylene oxide)-b-poly(ethylene oxide)-b-P[Asp(DET)] (P(EPE)-b-P[Asp(DET)]) used as an analog of P85-b-P[Asp(DET)], and pDNA were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service