Skip to Content
MilliporeSigma
All Photos(1)

Documents

P6030

Sigma-Aldrich

Potassium nitrate

suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

technique(s)

cell culture | mammalian: suitable

mp

334 °C (lit.)

solubility

H2O: 20/150 g/mL, clear, colorless

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentin Loux et al.
BMC genomics, 16, 296-296 (2015-04-18)
Propionibacterium freudenreichii (PF) is an actinobacterium used in cheese technology and for its probiotic properties. PF is also extremely adaptable to several ecological niches and can grow on a variety of carbon and nitrogen sources. The aim of this work
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
Mila Ivanova Luleva et al.
Sensors (Basel, Switzerland), 11(4), 4188-4206 (2011-12-14)
Displacement of soil particles caused by erosion influences soil condition and fertility. To date, the cesium 137 isotope ((137)Cs) technique is most commonly used for soil particle tracing. However when large areas are considered, the expensive soil sampling and analysis
Rose A Devasia et al.
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service