Skip to Content
MilliporeSigma
All Photos(1)

Documents

P183

Sigma-Aldrich

(+)-PD 128,907 hydrochloride

>97%, solid

Synonym(s):

(+)-(4aR,10bR)-3,4,4a,10b-Tetrahydro-4-propyl-2H,5H-[1]benzopyrano[4,3-b]-1,4-oxazin-9-ol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO3 · HCl
CAS Number:
Molecular Weight:
285.77
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

>97%

form

solid

color

off-white

solubility

ethanol: >10 mg/mL
DMSO: 18 mg/mL
H2O: 9 mg/mL

SMILES string

Cl.CCCN1CCO[C@H]2[C@H]1COc3ccc(O)cc23

InChI

1S/C14H19NO3.ClH/c1-2-5-15-6-7-17-14-11-8-10(16)3-4-13(11)18-9-12(14)15;/h3-4,8,12,14,16H,2,5-7,9H2,1H3;1H/t12-,14-;/m1./s1

InChI key

DCFXOTRONMKUJB-QMDUSEKHSA-N

Gene Information

human ... DRD3(1814)

Biochem/physiol Actions

Selective D3 dopamine receptor agonist.

Caution

Light sensitive

Legal Information

Manufactured and sold with the permission of Warner-Lambert Company.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A DeWald et al.
Journal of medicinal chemistry, 33(1), 445-450 (1990-01-01)
The dopamine agonist profile of (+-)-trans-3,4,4a,10b-tetrahydro-4-propyl-2H,5H-[1]benzopyrano [4,3-b]-1,4-oxazin-9-ol (16a) and its enantiomers (16b-c) was examined. Racemic 16a exhibited moderate affinity for the dopamine (DA) D2 receptor labeled with the DA antagonist ligand [3H]haloperidol and moderate in vivo activity; it attenuated gamma-butyrolactone-stimulated
F Sautel et al.
Neuroreport, 6(2), 329-332 (1995-01-26)
The functional potency of a series of dopamine agonists for increasing mitogenesis, measured by incorporation of [3H]thymidine, was established in transfected cell lines expressing human D2 or D3 receptors. The functional selectivity of agonists markedly differs from their binding selectivity.
Assessment of cocaine-like discriminative stimulus effects of dopamine D3 receptor ligands.
Acri, et al.
European Journal of Pharmacology, 281 (1995)
Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service