Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

O9506

Sigma-Aldrich

Oleic acid N-hydroxysuccinimide ester

~95% (TLC)

Synonym(s):

Succinimidyl oleate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$136.00
5 G
$497.00

$136.00


Estimated to ship onApril 19, 2025



Select a Size

Change View
1 G
$136.00
5 G
$497.00

About This Item

Empirical Formula (Hill Notation):
C22H37NO4
CAS Number:
Molecular Weight:
379.53
MDL number:
UNSPSC Code:
12352211

$136.00


Estimated to ship onApril 19, 2025


Assay

~95% (TLC)

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)ON1C(=O)CCC1=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified aminoacyl-tRNA. II. A general method for the preparation of acylaminoacyl-tRNA.
Y Lapidot et al.
Biochimica et biophysica acta, 145(2), 292-299 (1967-09-26)
A Al-Arif et al.
Journal of lipid research, 10(3), 344-345 (1969-05-01)
N-Hydroxysuccinimide esters of long-chain fatty acids have been used to synthesize the CoA and thioglycolic acid thiol esters of palmitic and 3-ketopalmitic acids in high yield and with a minimum of untoward side reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service