Skip to Content
MilliporeSigma
All Photos(1)

Documents

N8908

Sigma-Aldrich

N-(NBD-Aminocaproyl)sphingosine β-D-lactosyl

≥98%

Synonym(s):

C6-NBD Lactosyl cerebroside, N-[6-[(7-Nitro-2-1,3-benzoxadiazol-4-yl)amino]caproyl]-D-lactosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H69N5O16
Molecular Weight:
900.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98%

form

solid

storage temp.

−20°C

General description

Fluorescently labeled fatty acid sphingolipid membrane probe. NBD group excitation 460 nm, emission 534 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service