Skip to Content
MilliporeSigma
All Photos(1)

Documents

N3143

Sigma-Aldrich

Nalidixic acid sodium salt

powder

Synonym(s):

1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N2NaO3
CAS Number:
Molecular Weight:
254.22
Beilstein:
3580062
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

sterility

γ-irradiated

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

[Na+].CCN1C=C(C([O-])=O)C(=O)c2ccc(C)nc12

InChI

1S/C12H12N2O3.Na/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14;/h4-6H,3H2,1-2H3,(H,16,17);/q;+1/p-1

InChI key

ROKRAUFZFDQWLE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Recommended for use in molecular biology applications at 15 μg/ml

Biochem/physiol Actions

Nalidixic acid sodium salt is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. Inhibits nucleic acid and protein synthesis in Saccharomyces cerevisiae.

Reconstitution

Prepare stock solutions directly in the vial with sterile water (5-50 mg/ml). Stock solutions should be stored at −20 °C.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Duse et al.
Journal of dairy science, 98(9), 6387-6398 (2015-07-21)
Quinolone resistance may emerge in gut bacteria (e.g., in Escherichia coli) of animals. Such bacteria could cause infections in the animal itself or be transmitted to humans via the food chain. Quinolone resistance is also observed in fecal E. coli
Xiangqun Fan et al.
Environmental science and pollution research international, 20(6), 3649-3656 (2012-11-07)
The Fenton-like degradation of nalidixic acid was studied in this work. The effects of Fe(3+) concentration and initial H(2)O(2) concentration were investigated. Increasing the initial H(2)O(2) concentration enhances the degradation and mineralization efficiency for nalidixic acid, while Fe(3+) shows an
Mohamed Nawaz et al.
International journal of food microbiology, 155(3), 179-184 (2012-03-13)
A study was undertaken to isolate and characterize tetracycline and nalidixic acid-resistant Klebsiella spp. in farm-raised, imported shrimp sold in the United States. Sixty-seven multiple antibiotic-resistant Klebsiella spp. strains were isolated from imported shrimp samples. Using morphological and biochemical methods
Jing Wang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(11), 14631-14644 (2020-09-13)
During spaceflight, astronauts are subjected to various physical stressors including microgravity, which could cause immune dysfunction and thus potentially predispose astronauts to infections and illness. However, the mechanisms by which microgravity affects innate immunity remain largely unclear. In this study
Atefeh Abouzarzadeh et al.
Journal of molecular recognition : JMR, 25(7), 404-413 (2012-06-27)
For the first time in this work, uniform molecularly imprinted polymer (MIP) nanoparticles were prepared using nalidixic acid as a template. The MIP nanoparticles were successfully synthesized by precipitation polymerization applying methacrylic acid (MAA) as a functional monomer and trimethylolpropane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service