Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7445

Sigma-Aldrich

MNS

≥98% (HPLC), solid

Synonym(s):

3,4-Methylenedioxy-beta-nitrostyrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO4
CAS Number:
Molecular Weight:
193.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

yellow to green

solubility

H2O: <2 mg/mL
DMSO: ≥5 mg/mL

SMILES string

[O-][N+](=O)\C=C\c1ccc2OCOc2c1

InChI

1S/C9H7NO4/c11-10(12)4-3-7-1-2-8-9(5-7)14-6-13-8/h1-5H,6H2/b4-3+

InChI key

KFLWBZPSJQPRDD-ONEGZZNKSA-N

Biochem/physiol Actions

Src and Syk kinase inhibitor that prevents phosphorylation and cytoskeletal association of GPIIb/IIIa and talin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yangyang Liu et al.
Behavior genetics (2018-06-07)
The extent to which correlations between personality domains and physical and psychological health generalize cross-culturally is unclear. We compared the strength of associations between the personality domains and somatic and psychological distress in Chinese (N = 2069) and a genetically informative sample
Alexia E Metz et al.
Brain sciences, 9(2) (2019-02-10)
We examined the behavioral response (BR) and threshold (T) axes of Dunn's four-quadrant model of sensory processing (1997). We assessed whether they are ordinal ranges and if variation is associated with other similarly described characteristics: Introversion/Extraversion (I/E) of Eysenck's personality
Laura McGeown et al.
Behavioural brain research, 338, 66-75 (2017-10-17)
The social modeling of eating effect refers to the consistently demonstrated phenomenon that individuals tend to match their quantity of food intake to their eating companion. The current study sought to explore whether activity within the mirror neuron system (MNS)
Wei-Ya Wang et al.
Molecular pharmacology, 70(4), 1380-1389 (2006-07-14)
Binding fibrinogen to activated glycoprotein (GP)IIb/IIIa is the final common pathway of platelet aggregation and has become a successful target for antiplatelet therapy. In the present study, we found that a small chemical compound, 3,4-methyl-enedioxy-beta-nitrostyrene (MNS), exhibited potent and broad-spectrum
Ajit Gorakshakar et al.
The Indian journal of medical research, 146(3), 305-315 (2018-01-23)
The molecular basis of the blood group antigens was identified first in the 1980s and 1990s. Since then the importance of molecular biology in transfusion medicine has been described extensively by several investigators. Molecular genotyping of blood group antigens is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service