Skip to Content
MilliporeSigma
All Photos(2)

Documents

I9770

Supelco

Methyl indole-3-acetate

analytical standard, suitable for (for IAA Immunoassay Kit, PGR-3)

Synonym(s):

Indole-3-acetic acid methyl ester, Methyl 3-indolylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO2
CAS Number:
Molecular Weight:
189.21
Beilstein:
158031
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

analytical standard

Assay

≥99%

suitability

suitable for (for IAA Immunoassay Kit, PGR-3)

application(s)

agriculture

storage temp.

2-8°C

SMILES string

COC(=O)Cc1c[nH]c2ccccc12

InChI

1S/C11H11NO2/c1-14-11(13)6-8-7-12-10-5-3-2-4-9(8)10/h2-5,7,12H,6H2,1H3

InChI key

KTHADMDGDNYQRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian-Yu Mao et al.
Physiologia plantarum, 170(3), 318-334 (2020-08-06)
Weeping is a specific plant architecture with high ornamental value. Despite the considerable importance of the weeping habit to landscaping applications and knowledge of plant architecture biology, little is known regarding the underlying molecular mechanisms. In this study, growth and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service