Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I4397

Supelco

Hydrogen ionophore IV

Synonym(s):

ETH 1778, Octadecyl isonicotinate, Proton ionophore IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H41NO2
CAS Number:
Molecular Weight:
375.59
Beilstein:
5969390
MDL number:
UNSPSC Code:
26111700

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)c1ccncc1

InChI

1S/C24H41NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22-27-24(26)23-18-20-25-21-19-23/h18-21H,2-17,22H2,1H3

InChI key

VXDJNEIVBJLMMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service