Skip to Content
MilliporeSigma
All Photos(1)

Documents

I0654

Sigma-Aldrich

Interleukin-1 Soluble Receptor, Type I human

>97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

(IL-1 sRI)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>97% (SDS-PAGE)

form

lyophilized powder

mol wt

36 kDa

packaging

pkg of 0.1 mg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL1R1(3554)

General description

Interleukin-1 type 1 receptor (IL1R1) is a glycoprotein. It is mainly expressed on endothelial cells, epithelial cells, hepatocytes, fibroblasts, epidermal dendritic cells as well as T lymphocytes.

Biochem/physiol Actions

Interleukin-1 (IL-1), a cytokine, is a key regulator of acute and chronic inflammation. The IL-1 associated responses are achieved by association of IL-1 with IL-1 type 1 receptor (IL1R1).
Recombinant human IL-1 soluble receptor I is a transmembrane protein and a member of the immunoglobulin superfamily. It is expressed predominantly by T cells, fibroblasts, and endothelial cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5.0 mg bovine serum albumin

Analysis Note

The bioactivity is measured by its ability to inhibit the IL-1β-dependent proliferation of a murine helper T cell line, D10.G4.1.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F F Vasilyev et al.
Cellular & molecular immunology, 12(2), 222-230 (2014-07-01)
The biological activity of the multifunctional cytokine interleukin-1 (IL-1) is mediated by its receptors. The aim of this study was to determine if an association exists between single nucleotide polymorphisms (SNPs) in the IL-1 type 1 and 2 receptor genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service