Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA020026

Sigma-Aldrich

Anti-PDLIM5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Enigma homolog, Anti-Enigma-like PDZ and LIM domains protein, Anti-PDZ and LIM domain protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

TTQSRSFRILAQITGTEHLKESEADNTKKANNSQEPSPQLASSVASTRSMPESLDSPTSGRPGVTSLTTAAAFKPVGSTGVIKSPSWQRPNQGVPSTGRISNSATYSGSVAPANSALGQTQPSDQDTLVQRAEHIPAGKRTPMC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDLIM5(10611)

General description

PDLIM5 (PDZ and LIM domain 5) is a LIM domain containing protein consisting of an N-terminal PDZ (post-synaptic density-95/discs large/zone occludens-1) domain and three C-terminal LIM (Lin-11 Isl-1 mec03) domains. It is ubiquitously expressed in the brain. Cysteine-rich LIM domains are double zinc fingers type, which participates in the protein-protein interactions.

Immunogen

PDZ and LIM domain protein 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PDLIM5 (PDZ and LIM domain 5) is involved in the several cellular processes such as cytoskeleton organization, cell lineage specification, organ development, and oncogenesis. It also acts as a scaffold protein that helps to link protein kinases and transcription factors with cytoskeletal elements. Nonsynonymous polymorphisms of the PDLIM5 gene cause both bipolar disorder (BPD) and schizophrenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73770

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohd A Zain et al.
Psychiatric genetics, 23(6), 258-261 (2013-09-26)
Two single nucleotide polymorphisms of PDLIM5, rs7690296 and rs11097431, were genotyped using Mass-Array SNP genotyping by Sequenom technology in 244 bipolar disorder patients, 471 schizophrenia patients, and 601 control individuals who were Malay, Chinese, and Indian ethnic groups in the
Andrés D Maturana et al.
Cellular signalling, 23(5), 928-934 (2011-01-27)
Enigma homolog protein 1 (ENH1) acts as a scaffold that selectively associates protein kinases and transcription factors with cytoskeletal elements. ENH1 comprises an N-terminal PDZ domain and three C-terminal LIM domains. Through the LIM domains ENH1 interacts with the N-terminal
Mohd Aizat Zain et al.
BMC medical genetics, 13, 91-91 (2012-10-04)
One of the genes suggested to play an important role in the pathophysiology of bipolar disorder (BPD) is PDLIM5, which encodes LIM domain protein. Our main objective was to examine the effect of olanzapine treatment on PDLIM5 mRNA expression in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service