Documents
GETE70
GE TE70 ECL Transphor semi-dry transfer unit
gel size 14 cm (16 cm) , or 2 miniVue gels side by side, AC/DC input 100-240 V AC
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6210-34
Cytiva TE70
gel size
14 cm (16 cm) , or 2 miniVue gels side by side
Related Categories
General description
Designed to deliver outstanding electrophoretic transfer results using low current and voltage with a minimal amount of buffer. Units are available with and without built-in power supplies to give maximum flexibility.
  • •Electrotransfer proteins from polyacrylamide gels in less than 1 h in newly designed, state-of-the-art units.
  • •Tested for use with Amersham ECL Western blotting reagents
  • •Units are available with or without power supply.
  • •Use minimal buffer, only enough to saturate blotting papers and membranes.
  • •Long-lasting perforated platinum-titanium and stainless steel electrodes give uniform, contamination-free transfers.
  • •Transfer two polyacrylamide gels in a stack.
  • •Use low current and voltage for transfers.