Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE29-0588-06

Antibody package, Protein G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
NA.56

manufacturer/tradename

Cytiva

storage temp.

2-8°C

General description

Protein G is a good choice for general purpose capture of antibodies at laboratory scale since it binds a broader range of IgG from eukaryotic species and binds more classes of IgG than protein A.

Contents
HiTrap Protein G HP (1 × 1 ml)
HiTrap Desalting (1 × 5 ml)

The HiTrap Protein G HP column enclosed is recommended for most species of antibodies, including rat. A HiTrap Desalting column is also provided for quick pH neutralization of the eluted antibodies.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service