GE28-9035-29
TempliPhi™ Seq Resolver Kit
Cytiva 28-9035-29, sufficient for 20 reactions
Select a Size
$609.00
Select a Size
About This Item
$609.00
Recommended Products
form
liquid
usage
sufficient for 20 reactions
feature
hotstart: no
manufacturer/tradename
Cytiva 28-9035-29
color
colorless
shipped in
dry ice
storage temp.
−70°C
General description
Application
Features and Benefits
- For the amplification of difficult templates for successful DNA sequencing.
- Solve the most common sequencing problems such as repeats, sequencing stops, and compressions with a single kit.
- Improve difficult template sequencing success with up to 820 bp Phred 20 read length.
- Cultureless preparation from bacterial colonies, glycerol stock or purified DNA; saves time and reagents.
- Easy-to-use kit requires only 20 min hands-on time.
- Amplified DNA can be directly used in sequencing reactions.
- illustra™ TempliPhi™ Sequence Resolver Kit produces exceptional sequencing results from difficult templates such as those with high GC content and secondary structures.
- The kit resolves most of the sequencing stops or gaps that routinely elude other commonly used finishing methods.
Storage and Stability
Analysis Note
Legal Information
On packaging and insert for kits:
For use only as licensed by Qiagen GmbH. and Cytiva. The Phi 29 DNA polymerase may not be re-sold or used except in conjunction with the other components of this kit. See US patent numbers 5,854,033, 6,124,120, 6,143,495, 6,323,009, 5,198,543, 5,576,204, and their equivalents in other countries.
On container:
Subject to proprietary rights of Cytiva. May not be re-sold or used except with the other components of this kit. See US patent numbers 5,576,204, and their equivalents in other countries.
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service