Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE25-6400-80

TempliPhi Large Construct Kit

Cytiva 25-6400-80, sufficient for 1000 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
NACRES:
NA.85

usage

sufficient for 1000 reactions

manufacturer/tradename

Cytiva 25-6400-80

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

illustra TempliPhi Large Construct Kit. illustra TempliPhi Large Construct Kit uses isothermal rolling circle amplification (RCA) to efficiently prepare DNA sequencing templates specifically from large DNA vectors (e.g., fosmids and BACs).

Application

The TempliPhi Large Construct DNA Amplification Kit (TempliPhi LC Kit) was developed specifically to prepare templates for BAC or fosmid DNA sequencing. The TempliPhi method utilizes bacteriophage Phi29 DNA polymerase enzyme to exponentially amplify single- or double-stranded circular DNA templates by rolling circle amplification (RCA). This isothermal amplification method produces microgram quantities of DNA from picogram amounts of starting material in a few hours. Amplification in vitro of very small amounts of template DNA eliminates the need for overnight cell culture and conventional plasmid or M13 DNA purification. The proofreading activity of Phi29 DNA polymerase ensures high fidelity DNA replication.

The starting material for amplification can be a small amount of bacterial cells containing a BAC or fosmid DNA, or any circular DNA sample. Bacterial colonies can be picked from agar plates and added directly to the TempliPhi LC reaction. Alternatively, microliter quantities of a saturated bacterial culture or a glycerol stock can serve as starting material. Regardless of the source of starting material, amplification is completed in 18 h at 30°C with no needed for thermal cycling. The product of the TempliPhi Large Construct reaction is high molecular weight, double-stranded concatemers of the circular template. The TempliPhi Large Construct product is double-stranded DNA that can be sequenced with forward and reverse primers. DNA amplified by the TempliPhi Large Construct Amplification method can be used directly in cycle sequencing reactions without any purification.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
S-re the kit at -70°C. The enzyme mix must be s-red at -70°C; all other components may be s-red at -20°C Thaw components on ice and maintain at 0°C - 4°C during handling

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

TempliPhi
On packaging and insert for kits:
For use only as licensed by Qiagen GmbH. and Cytiva. The Phi 29 DNA polymerase may not be re-sold or used except in conjunction with the other components of this kit. See US patent numbers 5,854,033, 6,124,120, 6,143,495, 6,323,009, 5,198,543, 5,576,204, and their equivalents in other countries.
On container:
Subject to proprietary rights of Cytiva. May not be re-sold or used except with the other components of this kit. See US patent numbers 5,576,204, and their equivalents in other countries.
TempliPhi is a trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service