Skip to Content
MilliporeSigma
All Photos(2)

Documents

G5134

Sigma-Aldrich

α-D-Galactosamine 1-phosphate

~95%, powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14NO8P
CAS Number:
Molecular Weight:
259.15
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

~95%

form

powder

color

white

storage temp.

−20°C

SMILES string

O.NC1C(O)C(O)C(CO)OC1OP(O)(O)=O

InChI

1S/C6H14NO8P.H2O/c7-3-5(10)4(9)2(1-8)14-6(3)15-16(11,12)13;/h2-6,8-10H,1,7H2,(H2,11,12,13);1H2

InChI key

KPOAUMIAJAOYLX-UHFFFAOYSA-N

Biochem/physiol Actions

Galactosamine 1-phosphate and 3[H]UDP-glucose was used in the generation of radiolabeled UDP-GalNAc in the uridine moiety.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service