Skip to Content
MilliporeSigma
All Photos(1)

Documents

G0921

Sigma-Aldrich

6′-Guanidinonaltrindole ditrifluoroacetate dihydrate

≥97.5% (HPLC), solid

Synonym(s):

6′-GNTI, [(4bS,8R,8aS,14bR)-7-(Cyclopropylmethyl)-5,6,7,8,8a,9,14,14b-octahydro-1,8a-dihydroxy-4,8-methanobenzofuro[2,3-a]pyrido[4,3-b]carbazol-12-yl]-guanidine ditrifluoroacetate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29N5O3 · 2CF3CO2H · 2H2O
CAS Number:
Molecular Weight:
735.63
MDL number:
UNSPSC Code:
12352200

Assay

≥97.5% (HPLC)

form

solid

storage condition

desiccated
under inert gas

color

pink to tan

solubility

H2O: >20 mg/mL

storage temp.

−20°C

SMILES string

O.O.OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.NC(=N)Nc1ccc2c3C[C@@]4(O)[C@H]5Cc6ccc(O)c7OC(c3[nH]c2c1)[C@]4(CCN5CC8CC8)c67

Biochem/physiol Actions

6′-GNTI is a selective agonist of κ/δ opioid receptor heterodimers in the spinal cord and has the unique property of selectively activating only opioid receptor heterodimers and not homomers. 6′-GNT functions as an analgesic demonstrating that opioid receptor heterodimers are functionally relevant in vivo.

Caution

Air-sensitive

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service