Skip to Content
MilliporeSigma
All Photos(1)

Documents

F9553

Sigma-Aldrich

FTI-276 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

N-[4-[2-(R)-Amino-3-mercaptopropyl]amino-2-phenylbenzoyl]methionine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O3S2 ·C2HF3O2
CAS Number:
Molecular Weight:
547.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

film

storage condition

desiccated
under inert gas

color

off-white

solubility

H2O: >5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CSCC[C@H](NC(=O)c1ccc(NC[C@@H](N)CS)cc1-c2ccccc2)C(O)=O

InChI

1S/C21H27N3O3S2.C2HF3O2/c1-29-10-9-19(21(26)27)24-20(25)17-8-7-16(23-12-15(22)13-28)11-18(17)14-5-3-2-4-6-14;3-2(4,5)1(6)7/h2-8,11,15,19,23,28H,9-10,12-13,22H2,1H3,(H,24,25)(H,26,27);(H,6,7)/t15-,19+;/m1./s1

InChI key

IAVHISGQCODLHL-WSCVZUBPSA-N

Biochem/physiol Actions

FTI-276 is a highly potent RasCAAX peptidomimetic. FTI-276 antagonizes both H and K-Ras oncogenic signaling. It is an inhibitor of farnesyltransferase (Ftase) in vitro with an IC50 of 500 pM and is an anti-cancer agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michela Terlizzi et al.
Cells, 11(16) (2022-08-27)
Sphingosine-1-phosphate (S1P) is involved in inflammatory signaling/s associated with the development of respiratory disorders, including cancer. However, the underlying mechanism/s are still elusive. The aim of this study was to investigate the role of S1P on circulating blood cells obtained
Manpreet Malhi et al.
Communications biology, 4(1), 1239-1239 (2021-10-31)
Respiratory syncytial virus (RSV) is a leading cause of severe respiratory tract infections in children. To uncover new antiviral therapies, we developed a live cell-based high content screening approach for rapid identification of RSV inhibitors and characterized five drug classes
Xue Chen et al.
eLife, 10 (2021-02-03)
A farnesylated and methylated form of prelamin A called progerin causes Hutchinson-Gilford progeria syndrome (HGPS). Inhibiting progerin methylation by inactivating the isoprenylcysteine carboxylmethyltransferase (ICMT) gene stimulates proliferation of HGPS cells and improves survival of Zmpste24-deficient mice. However, we don't know

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service