Skip to Content
MilliporeSigma
All Photos(1)

Documents

F2917

Sigma-Aldrich

Fmoc-Nle-OH

Synonym(s):

(S)-2-(Fmoc-amino)caproic acid, (S)-2-(Fmoc-amino)hexanoic acid, N-(9-Fluorenylmethoxycarbonyl)-L-norleucine, Fmoc-L-norleucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Beilstein:
5305164
MDL number:
UNSPSC Code:
12352200

mp

141-144 °C (lit.)

storage temp.

−20°C

SMILES string

CCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4/c1-2-3-12-19(20(23)24)22-21(25)26-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,2-3,12-13H2,1H3,(H,22,25)(H,23,24)/t19-/m0/s1

InChI key

VCFCFPNRQDANPN-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service