Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU158661

Sigma-Aldrich

MISSION® esiRNA

targeting human NONO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAAATCTTCCTCCCGACATCACTGAGGAAGAAATGAGGAAACTATTTGAGAAATATGGAAAGGCAGGCGAAGTCTTCATTCATAAGGATAAAGGATTTGGCTTTATCCGCTTGGAAACCCGAACCCTAGCGGAGATTGCCAAAGTGGAGCTGGACAATATGCCACTCCGTGGAAAGCAGCTGCGTGTGCGCTTTGCCTGCCATAGTGCATCCCTTACAGTTCGAAACCTTCCTCAGTATGTGTCCAACGAACTGCTGGAAGAAGCCTTTTCTGTGTTTGGCCAGGTAGAGAGGGCTGTAGTCATTGTGGATGATCGAGGAAGGCCCTCAGGAAAAGGCATTGTTGAGTTCTCAGGGAAGCCAGCTGCTCGGAAAGCTCTGGACAGATGCAGTGAAGGCTCCTTCCTGCTAACCACATTTCCTCGTCCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Cheng et al.
Oncology reports, 39(6), 2575-2583 (2018-04-06)
Esophageal squamous cell carcinoma (ESCC) is one of the most common malignancies in China, and is associated with high morbidity and mortality. However, the molecular mechanisms that control ESCC tumorigenicity and metastasis remain unclear. Here, we report that the RNA
Taotao Han et al.
The Journal of clinical investigation, 130(7), 3901-3918 (2020-06-17)
Chronic infections can lead to carcinogenesis through inflammation-related mechanisms. Chronic infection of the human gastric mucosa with Helicobacter pylori is a well-known risk factor for gastric cancer. However, the mechanisms underlying H. pylori-induced gastric carcinogenesis are incompletely defined. We aimed
Lotte Victoria Winther Stagsted et al.
eLife, 10 (2021-01-22)
Circular RNAs (circRNAs) represent an abundant and conserved entity of non-coding RNAs; however, the principles of biogenesis are currently not fully understood. Here, we identify two factors, splicing factor proline/glutamine rich (SFPQ) and non-POU domain-containing octamer-binding protein (NONO), to be
Jarrod S Johnson et al.
Cell reports, 30(3), 914-931 (2020-01-23)
Transcriptional programming of the innate immune response is pivotal for host protection. However, the transcriptional mechanisms that link pathogen sensing with innate activation remain poorly understood. During HIV-1 infection, human dendritic cells (DCs) can detect the virus through an innate
Asma Chaoui et al.
Human molecular genetics, 24(17), 4933-4947 (2015-06-11)
SOX10 is a transcription factor with well-known functions in neural crest and oligodendrocyte development. Mutations in SOX10 were first associated with Waardenburg-Hirschsprung disease (WS4; deafness, pigmentation defects and intestinal aganglionosis). However, variable phenotypes that extend beyond the WS4 definition are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service