Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU135581

Sigma-Aldrich

MISSION® esiRNA

targeting human RUNX3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCTTCCCTCCTGTTCTCTGGTTATAGCTGGTCCCAGGTCAGCGTGGGAGGCACCTTTGGGTTCCCAGTGCCCAGCACTTTGTAGTCTCATCCCAGATTACTAACCCTTCCTGATCCTGGAGAGGCAGGGATAGTAAATAAATTGCTCTTCCTACCCCATCCCCCATCCCCTGACAAAAAGTGACGGCAGCCGTACTGAGTCTGTAAGGCCCAAAGTGGGTACAGACAGCCTGGGCTGGTAAAAGTAGGTCCTTATTTACAAGGCTGCGTTAAAGTTGTACTAGGCAAACACACTGATGTAGGAAGCACGAGGAAAGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jikui Sun et al.
Oncotarget, 8(67), 110785-110796 (2018-01-18)
Accumulating data demonstrates that the network dysregulation of microRNA-medicated target genes is involved in glioma. We have previously found miR-19a/b overexpression in glioma cell lines and specimens with various tumour grades. However, there was no report on the function and
J Justin Milner et al.
Nature, 552(7684), 253-257 (2017-12-07)
Tissue-resident memory CD8
Bufeng Zhuang et al.
Molecular medicine reports, 19(5), 3933-3940 (2019-03-01)
Dysregulated microRNAs (miRNAs/miRs) directly modulate the biological functions of non‑small cell lung cancer (NSCLC) cells and contribute to the initiation and progression of NSCLC; however, the specific roles and underlying mechanisms of the dysregulated miRNAs in NSCLC require further investigation.
Lin Shi et al.
Cell stress & chaperones, 25(5), 793-802 (2020-07-19)
Lung toxicity is the main cause of the death from methamphetamine (MA) abuse, but its mechanism has remained unclear. The purpose of our study was to investigate if MA can induce epithelial-to-mesenchymal transition (EMT) and if RUNX3 is involved in
Huaxia Chen et al.
Cell cycle (Georgetown, Tex.), 19(23), 3303-3316 (2020-11-03)
Keloid is an extremely common and often overlooked benign neoplastic disease, but its consequences should not be underestimated. Therefore, a deep exploration of the pathological mechanism of keloid becomes very essential. After 22 samples were collected from each patient's keloid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service