Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU047891

Sigma-Aldrich

MISSION® esiRNA

targeting human SIRT4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGAGCGTTTCCAAGTCCTGAACCCCACCTGGAGTGCTGAGGCCCATGGCCTGGCTCCTGATGGTGACGTCTTTCTCTCAGAGGAGCAAGTCCGGAGCTTTCAGGTCCCAACCTGCGTTCAATGTGGAGGCCATCTGAAACCAGATGTCGTTTTCTTCGGGGACACAGTGAACCCTGACAAGGTTGATTTTGTGCACAAGCGTGTAAAAGAAGCCGACTCCCTCTTGGTGGTGGGATCATCCTTGCAGGTATACTCTGGTTACAGGTTTATCCTCACTGCCTGGGAGAAGAAGCTCCCGATTGCAATACTGAACATTGGGCCCACACGGTCGGATGACTTGGCGTGTCTGAAACTGAATTCTCGTTGTGGAGAGTTGCTGCCTTTGATAGACCCATGCTGACCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongjie Sun et al.
OncoTargets and therapy, 11, 3959-3968 (2018-07-20)
Previous study has proven that SIRT4 is downregulated in gastric cancer (GC), but the role of SIRT4 has not been clearly understood. The aim of our work was to explore in detail the function and mechanism of SIRT4 in GC.
Ratana Lim et al.
Biology of reproduction, 95(5), 95-95 (2016-11-05)
Preterm birth remains the major cause of neonatal mortality and morbidity, mediated largely by an inflammatory process. The sirtuin (SIRT) family of cellular regulators has been implicated as key inhibitors of inflammation. We have previously reported a role for SIRT1
Peixin Huang et al.
Oncotarget, 6(13), 10812-10824 (2015-05-01)
Aging is the predominant risk factor for cardiovascular diseases and contributes to a considerably more severe outcome in patients with acute myocardial infarction. Resveratrol, a polyphenol found in red wine, is a caloric restriction mimetic with potential anti-aging properties which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service