Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E7528

Sigma-Aldrich

Anti-S1P5, C-Terminal antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Synonym(s):

Anti-EDG-8, Anti-Endothelial Cell Differentiation Gene 8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~42 kDa by SDS-PAGE

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... S1PR5(53637)

Immunogen

synthetic peptide (approximately 2.5 kDa, native) derived from the C-terminal region of the EDG-8/S1P5 receptor.

Application

Anti-S1P5, C-Terminal antibody produced in rabbit is suitable for western blotting at a working concentration of 5 to 10 μg/ml using RH7777 cells transfected with full length human EDG-8.

Biochem/physiol Actions

Sphingosine-1-phosphate receptor 5 (S1PR5) is a human gene which encodes a G protein-coupled receptor. It binds to lipid signaling molecule sphingosine 1-phosphate (S1P). S1PR5 regulates NK cell migration and distribution by interacting with sphingosine-1-phosphate (S1P). In brain endothelial cells (ECs), it contributes to optimal barrier formation and maintains immune quiescence of the barrier endothelium. S1PR5 is localized in the centrosome of mammalian cells and may participate in regulating mitosis. It may function as an intracellular target of S1P linked to regulation of mitosis.

Physical form

Supplied in phosphate buffered saline, pH 7.3, containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Xun Guan et al.
Zhongguo shi yan xue ye xue za zhi, 20(2), 412-415 (2012-05-01)
Natural killer (NK) cells can suppress the development of graft vs host disease (GVHD) while retaining antitumor response in allogeneic hematopoietic stem cell transplantation (allo-HSCT). Sphingosine-1-phosphate receptor 5 (S1PR5) can regulate NK cell migration and distribution in vivo by interacting
Ruben van Doorn et al.
Journal of neuroinflammation, 9, 133-133 (2012-06-22)
The sphingosine 1-phosphate (S1P) receptor modulator FTY720P (Gilenya®) potently reduces relapse rate and lesion activity in the neuroinflammatory disorder multiple sclerosis. Although most of its efficacy has been shown to be related to immunosuppression through the induction of lymphopenia, it
Laura Gillies et al.
Cellular signalling, 21(5), 675-684 (2009-02-13)
We show here that the endogenous sphingosine 1-phosphate 5 receptor (S1P(5), a G protein coupled receptor (GPCR) whose natural ligand is sphingosine 1-phosphate (S1P)) and sphingosine kinases 1 and 2 (SK1 and SK2), which catalyse formation of S1P, are co-localised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service