Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E3770

Sigma-Aldrich

rac-2-Ethoxy-3-octadecanamido-1-propylphosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H59N2O6P
CAS Number:
Molecular Weight:
550.75
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

impurities

≤0.5 mol/mol (NH4)2CO3

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)NCC(COP([O-])(=O)OCC[N+](C)(C)C)OCC

InChI

1S/C28H59N2O6P/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-28(31)29-25-27(34-7-2)26-36-37(32,33)35-24-23-30(3,4)5/h27H,6-26H2,1-5H3,(H-,29,31,32,33)

InChI key

QUVZKDZYEOYGKV-UHFFFAOYSA-N

Biochem/physiol Actions

Protein kinase C inhibitor.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Marx et al.
Journal of medicinal chemistry, 31(4), 858-863 (1988-04-01)
Previously unreported analogues of the synthetic antitumor phospholipid ET-18-OMe (1-octadecyl-2-methoxy-rac-glycero-3-phosphocholine), in which the 1-ether oxygen has been replaced by an amido group, have been prepared and evaluated for in vitro cytotoxic effects and for inhibition of protein kinase C. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service