Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D9671

Sigma-Aldrich

3,6-Diphenyl-1,2,4,5-tetrazine

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10N4
CAS Number:
Molecular Weight:
234.26
MDL number:
UNSPSC Code:
12352100

Assay

~98%

mp

198 °C (dec.) (lit.)

SMILES string

c1ccc(cc1)-c2nnc(nn2)-c3ccccc3

InChI

1S/C14H10N4/c1-3-7-11(8-4-1)13-15-17-14(18-16-13)12-9-5-2-6-10-12/h1-10H

InChI key

XAUWSIIGUUMHQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Wu Rao et al.
Bioorganic & medicinal chemistry letters, 16(14), 3702-3705 (2006-05-20)
Fourteen compounds of 3,6-disubstituted-1,4-dihydro-1,2,4,5-tetrazine derivatives were prepared and their structures were confirmed by single-crystal X-ray diffraction and the semi-empirical calculation of PM3 method. This reaction yields the 1,4-dihydro derivatives rather than the 1,2-dihydro derivatives. The central six-membered ring of 1,4-dihydro-1,2,4,5-tetrazine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service