CLS431413
Corning® glass fiber membrane filter discs
for the 1 L funnel, diam. 79 mm
Synonym(s):
filter accessory
Select a Size
$164.00
Select a Size
About This Item
$164.00
Recommended Products
Product Name
Corning® square glass fiber prefilter, for the 1 L funnel, diam. 79 mm
material
glass fiber membrane
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Corning 431413
diam.
79 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service