Recommended Products
product name
BE(2)-M17, 95011816
biological source
human nerve
growth mode
Adherent
karyotype
Not specified
morphology
Fusiform
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian neuroblastoma
Cell Line Description
The cell line BE(2)-M17 was isolated from SK-N-BE(2) (Sigma Catalogue number. 95011815) by limiting dilution. The cells show a population doubling time of 20-24 hours, grow in multi-layers but start to aggregate and float as the culture ages. The neuroblastic cells express properties of neuroadrenergic neurones and multiple, moderately long cell processes. BE(2)-M17 showed tyrosine hydroxylase and dopamine-ß-hydroxylase activity and specific uptake of norepinephrine (noradrenalin). Expression of intermediate filaments was detected as well as neuron-specific enolase and moderate expression of vimentin. BE(2)-M17 are tumorigenic in nude mice.
Application
Study of neuroblastoma
Culture Medium
EMEM (EBSS):Ham′s F12 (1:1) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS) (Heat Inactivated).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:8 to 1:20 i.e. seeding at 4x10,000 cells/cm2 using 0.1% trypsin or PBS only; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service