Skip to Content
MilliporeSigma
All Photos(1)

Documents

AFRC MAC 157

Reactivity: Anti-Cytokinin, plant, 93090997

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Hybridoma

ImmunoDon

F344 rat spleen

Myeloma

NS0

Reactivity

Anti-Cytokinin, plant

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS0 x Rat F344 spleen

Cell Line Description

The antibody is directed against the cytokinin isopentenyladenosine (2iPA) and related compounds in plant extracts.

Immunogen

Isopentenyladenosine

Application

Detection of 2iPA in plant tissue
The isotype of the monoclonal antibody produced by this cell line is rat IgG2a.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service