Skip to Content
MilliporeSigma
All Photos(1)

Documents

A10-85

93040117, rat nerve, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

A10-85, 93040117

biological source

rat nerve

growth mode

Adherent

karyotype

Modal no. 42

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat glioma

Cell Line Description

The glioma cell line A10-85 is a subclone of A10 cells which originally emerged from the parent line IA2 by reducing the serum concentration from 15% to 10%. This parent line IA2 was derived from a rat brain glioma induced transplacentally by ethylnitroso urea (inbred BDIxrats). A10-85 cells form processes in response to cAMP. GAPDH can be induced by hydrocortisone succinate and good growth was obtained in semi-solid medium. Injection of cells into syngeneric animals resulted in the formation of malignant astrocytic tumours after short latent periods.

Application

Experimental tumour models, carcinogenesis studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:50 to 1:100 seeding at 5x10,000 to 1x100,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service