Skip to Content
MilliporeSigma
All Photos(1)

Documents

M290

Reactivity: Anti-α-E (α-M290) integrin subunit, mouse, 93012001

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Hybridoma

ImmunoDon

LOU/c rat spleen

Myeloma

P3X63Ag8.653

Reactivity

Anti-α-E (α-M290) integrin subunit, mouse

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse P3X63Ag8.653 x Rat LOU/c spleen

Cell Line Description

No known cross reactivity. Blocks functional activity of mouse α-E-β-7 integrin.

Immunogen

Mouse gut intraepithelial lymphocytes

Application

Functional studies on α-M290 ß7 in mice
The isotype of the monoclonal antibody produced by this cell line is rat IgG2a.

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-5x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service