Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HL60 M2

88120803, human blood, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HL60 M2, 88120803

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian promyelocytic leukaemia

Cell Line Description

HL60 induced to mature into neutrophils with 1.75% DMSO after treating with SV40. Fails to express SV40 specific T Ag suggesting that HL60 M2 represents expansion of sub-clones of HL60 with inherent restricted capacity for neutrophil differentiation.

Application

Differentiation studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 1.25% Dimethyl Sulphoxide (DMSO).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. Do not allow the culture medium to become acidic or the cells will differentiate; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service