Skip to Content
MilliporeSigma
All Photos(1)

Documents

C1799

Sigma-Aldrich

Cytidine-2-14C 5′-triphosphate ammonium salt

≥95% (HPLC), aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H25N6O14P3
CAS Number:
Molecular Weight:
534.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

aqueous ethanol solution

extent of labeling

40-60 mCi per mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

N.NC1=NC(=O)N(C=C1)C2OC(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)C(O)C2O

Packaging

Combi-vial

Physical form

Aqueous solution containing 2% ethanol

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service