Skip to Content
MilliporeSigma
All Photos(1)

Documents

B2275

Sigma-Aldrich

N-Benzyl-9-(2-tetrahydropyranyl)adenine

suitable for plant cell culture

Synonym(s):

6-Benzylamino-9-(2-tetrahydropyranyl)-9H-purine, BPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N5O
CAS Number:
Molecular Weight:
309.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

technique(s)

cell culture | plant: suitable

storage temp.

−20°C

SMILES string

C1CCC(OC1)n2cnc3c(NCc4ccccc4)ncnc23

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Zhu et al.
Analytica chimica acta, 883, 81-89 (2015-06-20)
In the present work, a highly sensitive and selective biosensor based on aptamer-functionalized nanoporous gold film (NPGF) was successfully developed for direct electrochemical detection of bisphenol A (BPA). NPGF was prepared by dealloying Ag from Au/Ag alloy leaf in concentrated
John G Gibbons et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(8), 2485-2490 (2015-01-15)
Tandemly repeated ribosomal DNA (rDNA) arrays are among the most evolutionary dynamic loci of eukaryotic genomes. The loci code for essential cellular components, yet exhibit extensive copy number (CN) variation within and between species. CN might be partly determined by
Séverine H Collet et al.
Toxicology and applied pharmacology, 284(3), 323-329 (2015-03-12)
The investigation of interspecies differences in bisphenol A (BPA) pharmacokinetics (PK) may be useful for translating findings from animal studies to humans, identifying major processes involved in BPA clearance mechanisms, and predicting BPA PK parameters in man. For the first
Jillian Ashley-Martin et al.
Environmental research, 140, 360-368 (2015-04-29)
The fetal time period is a critical window of immune system development and resulting heightened susceptibility to the adverse effects of environmental exposures. Epidemiologists and toxicologists have hypothesized that phthalates, bisphenol A (BPA) and perfluoroalkyl substance have immunotoxic properties. Immunotoxic
Wei Wang et al.
Environment international, 83, 183-191 (2015-07-16)
Tetrabromobisphenol A (TBBPA) and eight bisphenol analogues (BPs) including bisphenol A (BPA) were determined in 388 indoor (including homes and microenvironments) dust samples collected from 12 countries (China, Colombia, Greece, India, Japan, Kuwait, Pakistan, Romania, Saudi Arabia, South Korea, U.S.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service