Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR96M2HSB

PCR amplification plate microplate

half skirt with a single notch, size 96 wells, blue, pkg of 5x10plates/cs

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells

material

conical bottom wells

feature

skirt

manufacturer/tradename

Axygen Inc PCR-96M2-HS-B
Corning

packaging

pkg of 5x10plates/cs

size

96 wells

color

blue

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service