Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV45694

Sigma-Aldrich

Anti-HAL antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-HIS, Anti-HSTD, Anti-Histidine ammonia-lyase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

72 kDa

species reactivity

mouse, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HAL(3034)

Immunogen

Synthetic peptide directed towards the C terminal region of human HAL

Application

Anti-HAL antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry of paraffin-embedded tissue sections at a concentration of 4-8μg/ml.

Biochem/physiol Actions

Histidine ammonia-lyase (HAL) is a cytosolic enzyme important in histidine catabolism. It catalyzes the nonoxidative deamination of L-histidine to trans-urocanic acid. Defective HAL activity results in accumulation of histidine and histamine in body fluids, a condition termed as hisdinemia.

Sequence

Synthetic peptide located within the following region: EAAHRLLLEQKVWEVAAPYIEKYRMEHIPESRPLSPTAFSLQFLHKKSTK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias Baedeker et al.
European journal of biochemistry, 269(6), 1790-1797 (2002-03-16)
Histidine ammonia-lyase (EC 4.3.1.3) catalyzes the nonoxidative elimination of the alpha-amino group of histidine using a 4-methylidene-imidazole-5-one (MIO), which is formed autocatalytically from the internal peptide segment 142Ala-Ser-Gly. The structure of the enzyme inhibited by a reaction with l-cysteine was
Yoko Kawai et al.
Human genetics, 116(5), 340-346 (2005-04-05)
Histidinemia (MIM235800) is characterized by elevated histidine in body fluids and decreased urocanic acid in blood and skin and results from histidase (histidine ammonia lyase, EC 4.3.1.3) deficiency. It is the most frequent inborn metabolic error in Japan. Although the
M Suchi et al.
Genomics, 29(1), 98-104 (1995-09-01)
Histidase (EC 4.3.1.3) is a cytosolic enzyme that catalyzes the nonoxidative deamination of histidine to urocanic acid. Histidinemia, resulting from reduced histidase activity as reported in Cambridge stock his/his mice and in humans, is the most frequent inborn metabolic error

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service