Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2009

Sigma-Aldrich

4-Monomethoxytrityl chloride

Synonym(s):

4-Methoxytriphenylchloromethane, 4-Methoxytrityl chloride, p-Anisylchlorodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17ClO
CAS Number:
Molecular Weight:
308.80
Beilstein:
798425
EC Number:
MDL number:
UNSPSC Code:
12352103

form

crystalline

color

off-white to pink

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(Cl)(c2ccccc2)c3ccccc3

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service