Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6317

Sigma-Aldrich

2′/3′-O-(N-Methylanthraniloyl)guanosine-5′-(γ-thio)triphosphate triethylammonium salt solution

≥90% (HPLC), aqueous solution

Synonym(s):

MANT-GTPγS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20N6O14P3S
Molecular Weight:
669.37
UNSPSC Code:
41106305
Pricing and availability is not currently available.

Assay

≥90% (HPLC)

form

aqueous solution

color

colorless

storage temp.

−20°C

Biochem/physiol Actions

Potent and competitive adenylyl cyclase inhibitor.
Potent and competitive adenylyl cyclase inhibitor. Competitively inhibited forskolin/Mn2+-stimulated adenylyl cyclase (AC) in S49 cyc- membranes (insect cells deficient in Gαs) with the IC50 value of 84 nM.

Other Notes

The product is a mixture of 2 esters.

Unit Definition

1 unit = 1 μl of 10 mM solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service