Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E8375

Sigma-Aldrich

Eserine

Synonym(s):

Physostigmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O2
CAS Number:
Molecular Weight:
275.35
Beilstein:
91230
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99% (HPLC)

mp

102-104 °C (lit.)

SMILES string

[H][C@]12N(C)CC[C@@]1(C)c3cc(OC(=O)NC)ccc3N2C

InChI

1S/C15H21N3O2/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2/h5-6,9,13H,7-8H2,1-4H3,(H,16,19)/t13-,15+/m1/s1

InChI key

PIJVFDBKTWXHHD-HIFRSBDPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Acetylcholinesterase inhibitor that crosses the blood-brain barrier and forms a carbamylated enzyme complex with acetyl cholinesterase that degrades slowly.
Acetylcholinesterase inhibitor that crosses the blood-brain barrier.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Nicotinic) and Acetylcholine Synthesis and Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ciaran G Carolan et al.
Journal of medicinal chemistry, 51(20), 6400-6409 (2008-09-27)
In this study, we report the SAR and characterization of two groups of isosorbide-based cholinesterase inhibitors. The first was based directly on the clinically used nitrate isosorbide mononitrate (ISMN) retention of the 5-nitrate group and introduction of a series of
Irina Esterlis et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(1), 78-82 (2012-11-20)
In vivo estimation of β(2)-nicotinic acetylcholine receptor availability with molecular neuroimaging is complicated by competition between the endogenous neurotransmitter acetylcholine and the radioligand (123)I-3-[2(S)-2-azetidinylmethoxy]pyridine ((123)I-5-IA). We examined whether binding of (123)I-5-IA is sensitive to increases in extracellular levels of acetylcholine
Federica Belluti et al.
Journal of medicinal chemistry, 48(13), 4444-4456 (2005-06-25)
In continuing research that led us to identify a new class of carbamate derivatives acting as potent (Rampa et al. J. Med. Chem. 1998, 41, 3976) and long-lasting (Rampa et al. J. Med. Chem. 2001, 44, 3810) acetylcholinesterase (AChE) inhibitors
Ciaran G Carolan et al.
Journal of medicinal chemistry, 53(3), 1190-1199 (2010-01-14)
Isosorbide-2-benzyl carbamate-5-benzoate is a highly potent and selective BuChE inhibitor. Meanwhile, isosorbide-2-aspirinate-5-salicylate is a highly effective aspirin prodrug that relies on the salicylate portion to interact productively with human BuChE. By integrating the salicylate group into the carbamate design, we
S J Reed et al.
British journal of anaesthesia, 110(4), 646-653 (2012-12-06)
Positron emission tomography studies in human subjects show that propofol-induced unconsciousness in humans is associated with a reduction in thalamic blood flow, suggesting that anaesthesia is associated with impairment of thalamic function. A recent study showed that antagonism of propofol-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service