Skip to Content
MilliporeSigma
All Photos(1)

Documents

E0779

Sigma-Aldrich

pBICEP-CMV-1 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

dry ice

storage temp.

−20°C

Application

For stable, cytoplasmic expression of N-terminal Met-FLAG fusion proteins from bicistronic mRNA. Stable expression is accomplished by neomycin selection (G 418 sulfate). Supplied with a pBICEP-CMV-1-lacZ Control Vector.

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pBICEP-CMV is a trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service