Skip to Content
MilliporeSigma
All Photos(1)

Documents

88535

Sigma-Aldrich

Thiocarbohydrazide

purum p.a., suitable for electron microscopy, ≥99.0% (N)

Synonym(s):

Thiocarbonyldihydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2NH)2CS
CAS Number:
Molecular Weight:
106.15
Beilstein:
506657
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum p.a.

Assay

≥99.0% (N)

form

crystals

technique(s)

electron microscopy: suitable

mp

171-174 °C (dec.) (lit.)

SMILES string

NNC(=S)NN

InChI

1S/CH6N4S/c2-4-1(6)5-3/h2-3H2,(H2,4,5,6)

InChI key

LJTFFORYSFGNCT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thiocarbohydrazide is used in silver proteinate specific and OTO-method (osmium-thiocarbohydrazide-osmium) methods for staining carbohydrates in electron microscopy. Thiocarbohydrazide may be used in the organic synthesis of metal ion coordinating compounds.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit Kumar Sharma et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(1), 337-342 (2010-11-26)
2,6-diacetyl pyridine based ligand was synthesized by the reaction of 2,6-diacetyl pyridine with thiocarbohydrazide in presence of acetic acid. The coordination compounds with Cr(III) and Ni(II) metal ions having [Cr(L)X]X2 and [Ni(L)X]X compositions (where L=ligand and X=NO3-, Cl- and CH3COO-)
Debabrata Maity et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(40), 11152-11161 (2011-09-02)
We present novel Schiff base ligands julolidine-carbonohydrazone 1 and julolidine-thiocarbonohydrazone 2 for selective detection of Cu(2+) in aqueous medium. The planar julolidine-based ligands can sense Cu(2+) colorimetrically with characteristic absorbance in the near-infrared (NIR, 700-1000 nm) region. Employing molecular probes
D Levanon et al.
The Histochemical journal, 31(1), 71-73 (1999-07-16)
Samples from seven sectors of the rabbit knee articular cartilage were shaved and prepared for the scanning electron microscope using either tannic acid, thiocarbohydrazide or nothing (control). Surface morphology was found to be more typical to a given sector and
D P Singh et al.
Journal of enzyme inhibition and medicinal chemistry, 22(2), 177-182 (2007-05-24)
A novel series of complexes of the type [M(TML)X2]; where TML is Tetradentate Macrocyclic Ligand; M = Co(II), Ni(II), Cu(II), Zn(II)or Cd(II); X = Cl-, CH3COO- or NO3- have been synthesized by template condensation of glyoxal and thiocarbohydrazide in the
Georg Kislinger et al.
iScience, 23(7), 101290-101290 (2020-07-06)
Volume electron microscopy enables the ultrastructural analysis of biological tissue. Currently, the techniques involving ultramicrotomy (ATUM, ssTEM) allow large fields of view but afford only limited z-resolution, whereas ion beam-milling approaches (FIB-SEM) yield isotropic voxels but are restricted in volume

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service