Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

78449

Sigma-Aldrich

o-Phenylenediamine dihydrochloride

tablet, ~30 mg substrate per tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3912045
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

tablet

mp

258 °C (dec.) (lit.)

solubility

H2O: 1 tablet/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

Phenylenediamine (O-Phenylenediamine) may be used in a wide variety of peroxidase based chromogenic assays and detection systems such as those based upon peroxidase conjugated primary and secondary antibodies.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    pH-tuned metal coordination and peroxidase activity of a peptide dendrimer enzyme model with a Fe(ii)bipyridine at its core.
    Geotti-Bianchini P, Darbre T, Reymond JL.
    Organic & Biomolecular Chemistry (2012)
    N Yokdang et al.
    British journal of cancer, 104(10), 1628-1640 (2011-04-21)
    Human breast carcinoma cells secrete an adenosine 5'-diphosphate transphosphorylase (sNDPK) known to induce endothelial cell tubulogenesis in a P2Y receptor-dependent manner. We examined sNDPK secretion and its effects on human endothelial cells. Nucleoside diphosphate kinase (NDPK) secretion was measured by
    Matthew A Wozniak et al.
    PloS one, 6(10), e25152-e25152 (2011-10-18)
    Alzheimer's disease (AD) afflicts around 20 million people worldwide and so there is an urgent need for effective treatment. Our research showing that herpes simplex virus type 1 (HSV1) is a risk factor for AD for the brains of people
    Cinzia Rizzo et al.
    Malaria journal, 10, 206-206 (2011-07-29)
    The Anopheles gambiae gSG6 is an anopheline-specific salivary protein which helps female mosquitoes to efficiently feed on blood. Besides its role in haematophagy, gSG6 is immunogenic and elicits in exposed individuals an IgG response, which may be used as indicator
    Heide Kogelberg et al.
    Journal of molecular biology, 382(2), 385-401 (2008-07-29)
    The alpha v beta 6 integrin is a promising target for cancer therapy. Its expression is up-regulated de novo on many types of carcinoma where it may activate transforming growth factor-beta1 and transforming growth factor-beta 3, interact with the specific

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service