Skip to Content
MilliporeSigma
All Photos(1)

Documents

77238

Sigma-Aldrich

N-Hexanoyl-D-sphingomyelin

≥98.0% (TLC)

Synonym(s):

(2S,3R,4E)-2-(Hexanoylamino)-4-octadecene-3-hydroxy-1-phosphocholine, N-(Hexanoyl)-sphing-4-enine-1-phosphocholine, N-Hexanoyl-D-erythro-sphingosylphosphorylcholine, C6 Sphingomyelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H59N2O6P
CAS Number:
Molecular Weight:
562.76
Beilstein:
18539759
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98.0% (TLC)

form

powder or solid

composition

carbon content, 61.89%
hydrogen content, 10.57%
nitrogen content, 4.98%

functional group

phospholipid

lipid type

sphingolipids

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C29H59N2O6P/c1-6-8-10-11-12-13-14-15-16-17-18-19-21-22-28(32)27(30-29(33)23-20-9-7-2)26-37-38(34,35)36-25-24-31(3,4)5/h21-22,27-28,32H,6-20,23-26H2,1-5H3,(H-,30,33,34,35)/b22-21+/t27-,28+/m0/s1

InChI key

AHZZHULAOVWYNO-RRKDMDGFSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service