Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76127

Sigma-Aldrich

2,3,3-Trimethyl-1-(4-sulfobutyl)indolium, inner salt

≥98.0% (HPLC)

Synonym(s):

1-(4-Sulfobutyl)-2,3,3-trimethylindolium, inner salt, 4-(2,3,3-Trimethylindolio)butanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO3S
CAS Number:
Molecular Weight:
295.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Product 76127 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥98.0% (HPLC)

form

powder

SMILES string

CC1=[N+](CCCCS([O-])(=O)=O)c2ccccc2C1(C)C

InChI

1S/C15H21NO3S/c1-12-15(2,3)13-8-4-5-9-14(13)16(12)10-6-7-11-20(17,18)19/h4-5,8-9H,6-7,10-11H2,1-3H3

InChI key

XTNXDHCAMPMVRG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service