Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

64010

Sigma-Aldrich

Metanil Yellow

for microscopy (Hist.), indicator (pH 1.2-2.3)

Synonym(s):

3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, Acid Yellow 36

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N3NaO3S
CAS Number:
Molecular Weight:
375.38
Colour Index Number:
13065
Beilstein:
3831568
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Hist.)
indicator (pH 1.2-2.3)

form

powder

solubility

methanol: 0.01 g/10 mL, orange to very deep orange

εmax

≥500 at 409-419  nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[O-]S(=O)(=O)c1cccc(c1)\N=N\c2ccc(Nc3ccccc3)cc2

InChI

1S/C18H15N3O3S.Na/c22-25(23,24)18-8-4-7-17(13-18)21-20-16-11-9-15(10-12-16)19-14-5-2-1-3-6-14;/h1-13,19H,(H,22,23,24);/q;+1/p-1/b21-20+;

InChI key

NYGZLYXAPMMJTE-ANVLNOONSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Metanil yellow has been used for staining of leukocyte granules. It is also used as a counterstain in microscopic sections.

Biochem/physiol Actions

Metanil yellow is an azo dye. It is used as a counterstain in PAS (periodic acid-Schiff)/iron-hematoxylin stained sections to increase contrast.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic degradation of azo dye Metanil Yellow: optimization and kinetic modeling using a chemometric approach.
Applied Catalysis. B, Environmental, 77, 1-11 (2007)
An improved stain for connective tissue cells in plastic-embedded sections.
Laboratory Medicine, 10, 783-785 (1979)
Enhancement of histological detail using metanil yellow as counterstain in periodic acid Schiff's hematoxylin staining of glycol methacrylate tissue sections.
Quintero-Hunter I, et al.
Biotechnic & Histochemistry, 66, 169-169 (1991)
Cellular localization of latent murine cytomegalovirus.
Koffron AJ, et al.
Journal of Virology, 72, 95-95 (1998)
P B Rastogi et al.
Journal of food protection, 59(3), 244-248 (1996-03-01)
Among a variety of experimental protocols used, the combined use of 0.5% dextrose in bottom agar and 1 micromol of flavin mononucleotide (FMN) in preincubation mixtures without fraction S9 mix resulted in the highest level of induced Salmonella typhimurium his

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service