Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55497

Sigma-Aldrich

Hydroxyapatite

′fast flow′

Synonym(s):

Calcium phosphate hydroxide, Durapatite, Hydroxylapatite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3Ca3(PO4)2 · Ca(OH)2
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

capacity

0.7 mg/g, dry material binding capacity (DNA)
12 mg/g, dry material binding capacity (BSA)

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Other Notes

Purification of proteins; Separation of single-stranded RNA from double-stranded RNA; Purification of DNA

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxyapatite column chromatography in procedures for isolation of purified DNA.
G G Markov et al.
Analytical biochemistry, 59(2), 555-563 (1974-06-01)
The DNA polymerases of KB cells.
W D Sedwick et al.
Methods in enzymology, 29, 89-102 (1974-01-01)
Preparation of tRNA's and aminoacyl-tRNA synthetases from Bacillus subtilis cells at various stages of growth and spores.
B S Vold
Methods in enzymology, 29, 502-510 (1974-01-01)
A simple method for the separation of single-stranded and double-stranded RNA on hydroxyapatite.
J Kalmakoff et al.
Analytical biochemistry, 55(1), 26-33 (1973-09-01)
N T Redpath et al.
European journal of biochemistry, 212(2), 511-520 (1993-03-01)
Eukaryotic elongation-factor-2 kinase has been purified to homogeneity from rabbit reticulocytes through a seven-step procedure and has been identified as a protein with a molecular mass of approximately 103 kDa as judged by SDS/PAGE. A degradation product of about 95

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service